Create stunning time-lapse images in your lab
Designed specifically for the EVOS M5000 Imaging System and EVOS M7000 Imaging System, the EVOS Onstage Incubator is an environmental chamber that enables precise control of temperature, humidity, and three gases for time-lapse imaging of live cells under both physiological and non-physiological conditions.
Request a demoLearn more about EVOS M7000
Application guide: Setting up hypoxic conditions with the EVOS onstage incubator. Download now
Superb flexibility, ease of use, and optical performance for live cell imaging
Environmental settings and image acquisition parameters are all seamlessly integrated into the EVOS M5000 and EVOS M7000 interfaces, creating a high-performance inverted imaging system with superb flexibility, ease of use and optical performance for demanding live cell time-lapse imaging experiments. Features include:
- Fully integrated environmental chamber for live cell time-lapse imaging
- Intuitively set environmental and image acquisition parameters
- Easily maintain physiological or non-physiological conditions with precision control
- Small footprint, robust build, and sleek design
- Choice of a range of vessel holders
Time-lapse videos using the EVOS FL Auto 2 with Onstage Incubator
Cell division in HeLa cells
Cells: HeLa
Reagents: CellLight H2B GFP, CellLight Mito RFP
Time Lapse: 24 hours with images taken every 12 minutes
Imaged on EVOS FL Auto Imaging System using the Onstage Incubator
Disruption of microfilament organization in HeLa cells
Cells: HeLa treated with Latrunculin A
Reagents: CellLight Actin GFP, MitoTracker Red, NucBlue Live
Time Lapse: 14 hours with images taken every 5 minutes
Imaged on EVOS FL Auto Imaging System using the Onstage Incubator
Specifications
| Attribute | Details |
|---|---|
| Compatible Vessels | Well plates, 35 mm dishes, T25 Flasks |
| Temperature Range | Ambient - 40oC |
| CO2 % Range | 0-20% |
| O2 % Range | Ambient - 0% |
| Humidity Range | >80% RH @ 37oC |
| Dimensions |
|
| Weight |
|
Resources
BioProbes Journal Article
Scientific Posters
Application Notes
Service
We provide a full range of service options for all EVOS cell imaging systems.
EVOS Accessories
EVOS Application Guides
For Research Use Only. Not for use in diagnostic procedures.