photo of the CellInsight CX7 High-Content Screening (HCS) Platform
The Thermo Scientific CellInsight CX7 High-Content Screening (HCS) Platform is an integrated system for all-around performance in screening and analysis.
Flexible imaging options with the CellInsight CX7 High-Content Screening Platform
With the CellInsight CX7 High-Content Screening Platform you have a choice of imaging modes to extract the information you need from your samples. Use the entire fluorescence spectrum to optimize your assay, and select either widefield or confocal optics for any channel. You also have 4-color brightfield options for colorimetric analysis of tissue sections. You can’t always predict what assays are coming next, but the CellInsight CX7 HCS Platform has you covered.
CellInsight CX7 system highlights
Illumination7-channel solid-state LED light engine
CameraPhotometrics High-Resolution Fluorescent Camera 
Widefield7-channel fluorescent imaging
Brightfield5-channel brightfield imaging
Confocal7-channel confocal imaging
Objectives3-position objective (range from 2x–60x, low and high NA)
FocusSoftware and laser-based autofocus for consistent scan times
AutomationConfigured for fully automated plate handling and scanning
SoftwareHCS Studio software for integrated data collection and analysis
HeLa cells imaged using the CellInsight CX7 platform
HeLa cell image acquired using the Thermo Scientific CellInsight CX7 platform and stained with Invitrogen HCS NuclearMask Blue (blue), Invitrogen MitoTracker Orange (green), and Invitrogen Alexa Fluor 647 Phalloidin (red).
A549 cells imaged using the CellInsight CX7 platform
A549 cell image acquired using the Thermo Scientific CellInsight CX7 platform and stained with Invitrogen HCS CellMask Blue (blue), Invitrogen Alexa Fluor 488 Phalloidin (green), and Invitrogen Alexa Fluor 750 goat anti–mouse IgG secondary antibody (pink).
Three-color brightfield image of a fixed tissue slice imaged with the CellInsight CX7 platform.
Three-color brightfield image of fixed human breast cancer tissue stained with KI67-DAB and counterstained with hematoxylin and eosin and imaged with the Thermo Scientific CellInsight CX7 platform.
Reagents optimized for high-content screening
Drawing on decades of experience in fluorescence imaging, Invitrogen HCS products are developed using Thermo Scientific High-Content Platforms with special considerations for the high-throughput workflow and automated imaging:
  • Cell and nuclear masks for automated demarcation
  • Robust functional probes for cell health interrogation
  • Flexible assay workflow for automated processing
  • Broad fluorophore choice for easy multiplexing
  • Validated on multiple imaging platforms
Select reagents for use with the CellInsight CX7
Other cell structure and segmentation tools
Clear identification of cell and nuclear boundaries in automated imaging requires fiducial markers that will label cellular structures robustly and reproducibly. Cell masks are designed specifically for automated imaging. Molecular Probes cell structure labels are widely used in microscope imaging and HCS.
See other reagents for cell structure
Other cell health and cytotoxicity assays
We offer a diverse selection of imaging and microscopy assays for the analysis of cell viability, proliferation, apoptosis, cell cycle and DNA repair, migration, adhesion and other cellular functions. Many of these assays have been validated specifically for HCS platforms.
Browse cell health and cytotoxicity assays by application:
Microplates and slides
Compatible with SBS standard microplates (6–1,536 well) and slides
Automation
Orbitor RS 2 Microplate Mover
Increase throughput, storage capacity and operational flexibility with the Thermo Scientific Orbitor RS 2 Microplate Mover—industrial-sized performance within a compact, benchtop mover.
The Orbitor RS 2 is a high-speed microplate mover offering proven, reliable performance with totally flexible plate handling. Extensive vertical reach allows multiple stacked or high density instruments to be loaded in a small footprint, and a bi-directional telescoping arm provides superior reach, improved user safety and unlimited base rotations within a 360° workspace.
CameraPhotometrics High-Resolution Fluorescent Camera:
  • High-resolution, thermoelectrically cooled to 0°C
  • 14-bit dynamic range
  • FOV 2208 x 2208 pixel array
  • 4.54 micron pixel size
  • 75% peak quantum efficiency (QE)
Data managementCompatible with Thermo Scientific Store Image and Database Management Software
Dimensions20 in. x 32 in. x 18 in. (50.8 cm x 81.3 cm x 45.7 cm)
Image compatibilityJPG, BMP, GIF, PNG, TIF, C01, DIB
Light sourceLED, solid-state 7-color light engine used with provided filter sets offers the following  excitation/emission capabilities (Ex/Em, in nm):
  • 386/440 (e.g., Alexa Fluor 350 dye)
  • 438/480 (e.g., Alexa Fluor 405 dye)
  • 485/521 (e.g., Alexa Fluor 488 dye)
  • 549/600 (e.g., Alexa Fluor 555 dye)
  • 560/607 (e.g., Alexa Fluor 594 dye)
  • 650/694 (e.g., Alexa Fluor 647 dye)
  • 740/810 (e.g., Alexa Fluor 750 dye)
ObjectivesStandard (Olympus™ objectives):
  • 10x (0.30NA)
  • 20x (0.4NA)
  • 20x (0.7 NA) Olympus objectives
  • 40x (0.6 NA) Olympus objectives
Optional (Olympus objectives):
  • 2x (0.08NA)
  • 4x (0.16NA)
PCWindows 7 Professional 64-Bit PC with 32GB (4x8GB) RAM, Intel® Xeon® Processor 3.7GHz Turbo, RAID I Controller with 1.8TB Hard Drive and 256 Boot Drive.
Includes Dell 24" High Resolution Widescreen Monitor or equivalent, keyboard and mouse.
SoftwareThermo Scientific HCS Studio 4.0 Cell Analysis Software
Wattage300 W
Weight150 lb (68 kg)
Reagents optimized for high-content screening
Drawing on decades of experience in fluorescence imaging, Invitrogen HCS products are developed using Thermo Scientific High-Content Platforms with special considerations for the high-throughput workflow and automated imaging:
  • Cell and nuclear masks for automated demarcation
  • Robust functional probes for cell health interrogation
  • Flexible assay workflow for automated processing
  • Broad fluorophore choice for easy multiplexing
  • Validated on multiple imaging platforms
Select reagents for use with the CellInsight CX7
Other cell structure and segmentation tools
Clear identification of cell and nuclear boundaries in automated imaging requires fiducial markers that will label cellular structures robustly and reproducibly. Cell masks are designed specifically for automated imaging. Molecular Probes cell structure labels are widely used in microscope imaging and HCS.
See other reagents for cell structure
Other cell health and cytotoxicity assays
We offer a diverse selection of imaging and microscopy assays for the analysis of cell viability, proliferation, apoptosis, cell cycle and DNA repair, migration, adhesion and other cellular functions. Many of these assays have been validated specifically for HCS platforms.
Browse cell health and cytotoxicity assays by application:
Microplates and slides
Compatible with SBS standard microplates (6–1,536 well) and slides
Automation
Orbitor RS 2 Microplate Mover
Increase throughput, storage capacity and operational flexibility with the Thermo Scientific Orbitor RS 2 Microplate Mover—industrial-sized performance within a compact, benchtop mover.
The Orbitor RS 2 is a high-speed microplate mover offering proven, reliable performance with totally flexible plate handling. Extensive vertical reach allows multiple stacked or high density instruments to be loaded in a small footprint, and a bi-directional telescoping arm provides superior reach, improved user safety and unlimited base rotations within a 360° workspace.
CameraPhotometrics High-Resolution Fluorescent Camera:
  • High-resolution, thermoelectrically cooled to 0°C
  • 14-bit dynamic range
  • FOV 2208 x 2208 pixel array
  • 4.54 micron pixel size
  • 75% peak quantum efficiency (QE)
Data managementCompatible with Thermo Scientific Store Image and Database Management Software
Dimensions20 in. x 32 in. x 18 in. (50.8 cm x 81.3 cm x 45.7 cm)
Image compatibilityJPG, BMP, GIF, PNG, TIF, C01, DIB
Light sourceLED, solid-state 7-color light engine used with provided filter sets offers the following  excitation/emission capabilities (Ex/Em, in nm):
  • 386/440 (e.g., Alexa Fluor 350 dye)
  • 438/480 (e.g., Alexa Fluor 405 dye)
  • 485/521 (e.g., Alexa Fluor 488 dye)
  • 549/600 (e.g., Alexa Fluor 555 dye)
  • 560/607 (e.g., Alexa Fluor 594 dye)
  • 650/694 (e.g., Alexa Fluor 647 dye)
  • 740/810 (e.g., Alexa Fluor 750 dye)
ObjectivesStandard (Olympus™ objectives):
  • 10x (0.30NA)
  • 20x (0.4NA)
  • 20x (0.7 NA) Olympus objectives
  • 40x (0.6 NA) Olympus objectives
Optional (Olympus objectives):
  • 2x (0.08NA)
  • 4x (0.16NA)
PCWindows 7 Professional 64-Bit PC with 32GB (4x8GB) RAM, Intel® Xeon® Processor 3.7GHz Turbo, RAID I Controller with 1.8TB Hard Drive and 256 Boot Drive.
Includes Dell 24" High Resolution Widescreen Monitor or equivalent, keyboard and mouse.
SoftwareThermo Scientific HCS Studio 4.0 Cell Analysis Software
Wattage300 W
Weight150 lb (68 kg)
Learn more about 2D/3D visualization and analysis (2D–5D)
With incredible speed and flexibility, Amira Software supports advanced 2D–5D bioimaging workflows in research areas ranging from structural and cellular biology to tissue imaging, neuroscience, preclinical imaging and bioengineering.
Learn more about Amira software